missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Benzoate, TRC
Specifications
Specifications
| Kemiskt namn eller material | Propyl Benzoate |
| CAS | 2315-68-6 |
| Molekylformel | C10 H12 O2 |
| Kvantitet | 1 g |
| Synonym | Propyl Benzoate, Benzoic acid','propyl ester, NSC 229333, Propyl benzoate, n-Propyl benzoate |
| InChI formel | InChI=1S/C10H12O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3 |
| LEDER | CCCOC(=O)c1ccccc1 |
| IUPAC-namn | propyl benzoate |
| Molekylvikt (g/mol) | 164.201 |
| Formel vikt | 164.0837 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?