missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl gallate, 98%, Thermo Scientific Chemicals
253.25 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 146.0°C to 150.0°C |
Color | White to Beige |
Loss on Drying | 0.5% max. (110°C, 4 hrs) |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Quantity | 100 g |
Merck Index | 15, 7971 |
Solubility Information | Solubility in water: 0.35g/100 mL (25°C). Other solubilities: 103g/100 g alcohol,83g/100 g ether,soluble in aceton,propyleenglycol and,polysorbaat 8 |
Formula Weight | 212.2 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Propyl gallate
- Warning
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'