Learn More
Propyl isothiocyanate, 97%, Thermo Scientific™
Catalog Number p-2704886
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
628-30-8
C4H7NS
101.167
MFCD00004823
KKASGUHLXWAKEZ-UHFFFAOYSA-N
propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
69403
1-isothiocyanatopropane
CCCN=C=S
628-30-8
C4H7NS
101.167
MFCD00004823
KKASGUHLXWAKEZ-UHFFFAOYSA-N
propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
69403
1-isothiocyanatopropane
CCCN=C=S
CAS Min % 97.0
Density 0.9780g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2CH2NCS
Quantity 1 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Propyl isothiocyanate