Läs mer
Propylene Glycol Dicaprate, TRC
Produktkod. 30424569
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30424569 100 mg
1 options
Produktkod. 30424569 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP797145100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propylene Glycol Dicaprate
CAS 53824-77-4
Molekylformel C23 H44 O4
Kvantitet 100 mg
Synonym Propylene Glycol Dicaprate, 1,2-Propanediol Didecanoate, Captex 100, Nikkol PDD, Propylene glycol didecanoate
InChI formel InChI=1S/C23H44O4/c1-4-6-8-10-12-14-16-18-22(24)26-20-21(3)27-23(25)19-17-15-13-11-9-7-5-2/h21H,4-20H2,1-3H3
LEDER CCCCCCCCCC(=O)OCC(C)OC(=O)CCCCCCCCC
IUPAC-namn 2-decanoyloxypropyl decanoate
Molekylvikt (g/mol) 384.593
Formel vikt 384.324