Läs mer
Propyliodone, LoGiCal
Produktkod. 30458056
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30458056 10 mg
1 options
Produktkod. 30458056 Leverantör LoGiCal Leverantörsnummer LGCFOR3199.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Propyliodone
CAS 587-61-1
Analyt- eller komponentnamn Propyliodone
Frakt skick Room Temperature
Molekylformel C10 H11 I2 N O3
Certifikat och efterlevnad ISO 9001
Synonym 1(4H)-Pyridineacetic acid, 3,5-diiodo-4-oxo-, propyl ester, 3,5-Diiodo-4-oxo-1(4H)-pyridineacetic acid propyl ester, 3,5-Diiodo-4-pyridone-N-acetic acid propyl ester, A. G. 33-107, Bronchodiagnostin, Brosombra, Dionosil, Diostril, NSC 97103, Propiodone, Propyl (3,5-diiodo-4-oxo-1,4-dihydro-1-pyridyl)acetate, Propyl 3,5-diiodo-4-pyridone-N-acetate, Propyliodon, Propyliodone
InChI formel InChI=1S/C10H11I2NO3/c1-2-3-16-9(14)6-13-4-7(11)10(15)8(12)5-13/h4-5H,2-3,6H2,1H3
Lösningstyp Neat
LEDER CCCOC(=O)CN1C=C(I)C(=O)C(=C1)I
IUPAC-namn propyl 2-(3,5-diiodo-4-oxopyridin-1-yl)acetate
Industry Type Clinical Testing
Molekylvikt (g/mol) 447.01
Kvantitet 10 mg
Formel vikt 446.8828 g/mol