Learn More
Prostaglandin E1, 98+%, Thermo Scientific Chemicals
Catalog Number 10592441
5775.16 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 mg
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 745-65-3
Melting Point 115.0°C to 116.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H34O5
MDL Number MFCD00077860
Quantity 10 mg
Merck Index 15, 7987
Specific Rotation Condition −64.00 (20.00°C c=1.0,C2H5OH)
Specific Rotation -64
Synonym alprostadil, prostaglandin e1, pge1, muse, alprostadilum, prostavasin, topiglan, vasaprostan, femprox, edex
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,methanol,ethyl acetate and,acetone
InChI Key GMVPRGQOIOIIMI-DWKJAMRDSA-N
SMILES CCCCCC(C=CC1C(CC(=O)C1CCCCCCC(=O)O)O)O
IUPAC Name 7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]heptanoic acid
Molecular Weight (g/mol) 354.48
PubChem CID 5280723
ChEBI CHEBI:15544
Formula Weight 354.48
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Prostaglandin E1, 98+%