Läs mer
Prosulfuron, TRC
Produktkod. 17963195
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17963195 100 mg
17993595 1 g
17951815 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17963195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP838688100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94125-34-5
Molekylformel C15 H16 F3 N5 O4 S
Molekylvikt (g/mol) 419.38
Synonym N-[[(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]-2-(3,3,3-trifluoropropyl)benzenesulfonamide, CGA 152005, N-(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)-N'-[2-(3,3,3-trifluoroprop-1-yl)benzenesulfonyl]urea, Peak, Peak 57WG, Pik, Prosulfuron
IUPAC-namn 1-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-3-[2-(3,3,3-trifluoropropyl)phenyl]sulfonylurea
LEDER COc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2CCC(F)(F)F)n1
Specifikationer
Kemiskt namn eller material Prosulfuron
Kvantitet 100 mg
InChI formel InChI=1S/C15H16F3N5O4S/c1-9-19-12(22-14(20-9)27-2)21-13(24)23-28(25,26)11-6-4-3-5-10(11)7-8-15(16,17)18/h3-6H,7-8H2,1-2H3,(H2,19,20,21,22,23,24)
Formel vikt 419.0875