Learn More
Thermo Scientific™ ProSwift™ ConA-1S Concanavalin A Purification of Con A Binding Glycans and Glycoconjugates HPLC Column, Monolith, 5 mm x 50 mm
Column Format: Analytical Column; Pore Size: Monolith; Column Phase: Affinity Separation
Brand: Thermo Scientific™ 074148
Additional Details : Weight : 0.50000kg
Description
For fast, highly efficient enrichment and purification of Con A binding glycans and glycoconjugates, choose the Thermo Scientific™ ProSwift™ ConA-1S LC Column. With the high capacity and ligand density of this monolith column, you can achieve highly efficient enrichment of samples and purify up to 2mg of glycoproteins. The high peak efficiency produces sharp, narrow peaks, resulting in low elution volumes of highly concentrated products. Designed for use in HPLC systems, the ProSwift ConA-1S LC column provides many advantages compared to standard manual methods.
Excellent Performance in HPLC Systems:- Faster separations, better enrichment and higher sample recovery
- Automatable analysis and high-throughput capability
- More accurate analysis with on-line monitoring
- Reusable for more than 100 purifications with minimal loss of capability
- Maintains specificity and selectivity for ConA binding glycans and glycoconjugates
Specifications
Analytical Column | |
2 mL/min | |
50 mm | |
Monolith | |
Monolith | |
30°C | |
Monolith | |
Purification of Con A Binding Glycans and Glycoconjugates | |
Analytical Column |
5 mm | |
HPLC | |
2000 psi (140 bar) | |
5 to 8 | |
Up to 10% Methanol | |
Concanavalin A | |
Monolith Polymer | |
Affinity Separation |