Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Ferrules
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Ferrules offer complete versatility for connecting capillary tubing.
Brand: Trajan Scientific and Medical™ 213007
Additional Details : Weight : 0.01000kg
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
1.59 mm to 1/32 in. | |
ProteCol PEEK Ferrules 1/16 in. to 1/32 in. |