Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Fingertight Fittings
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Fingertight Fittings offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 222002
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15249213
  • 520.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
PEEK Fingertight Fittings 1/32 in. to 10-32UNF
Stainless steel
1/32 in. to 10-32UNF
Videos