Learn More
Trajan Scientific and Medical™ ProteCol™ Stainless Steel Unions
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ Stainless Steel Unions offer complete versatility for connecting capillary tubing.
Brand: Trajan Scientific and Medical™ 211010
Additional Details : Weight : 0.01000kg
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
Union | |
Size: 1/16 in. to 1/16 in. only union |