missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Protein A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB99761-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Protein A Monoclonal specifically detects Protein A in Multi-Species samples. It is validated for Western Blot.
Specifikationer
| Protein A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects recombinant Protein A in direct ELISA and Western blots. | |
| All species | |
| Supernatant |
| Western Blot | |
| 946418 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
| Immunoglobulin G binding protein A, PROA, Protein-A, SPA, Staphylococcal protein A | |
| Recombinant Protein A | |
| 100 μg | |
| 3919448.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering