Learn More
Abeba™ Protektor Line 1655 Shoes
Leather upper construction includes reflective material.
Brand: Abeba™ 1655/44
Additional Details : Weight : 1.32000kg
Description
- CE EN ISO 20345:2011 S3 SRC
- Smooth leather
- Black
- Steel toe cap
- Penetration resistant steel midsole
- Slip resistance PU outsole sole
- Antistatic
- Leather lining
- velcro fastening
- Reflective material
Specifications
CE EN ISO 20345:2011 S3 SRC | |
Leather | |
Slip-resistant | |
1655 |
Black | |
44 | |
Polyurethane | |
Steel |