Läs mer
Prothiofos, TRC
Produktkod. 30412971
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30412971 100 mg
1 options
Produktkod. 30412971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP838840100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Prothiophos
CAS 34643-46-4
Molekylformel C11 H15 Cl2 O2 P S2
Kvantitet 100 mg
Synonym Phosphorodithioic acid','O-(2,4-dichlorophenyl) O-ethyl S-propyl ester, BAY-NTN 8629, Dichlorpropaphos, Prothiofos, Prothiophos, Tokuthion, Toyodan
InChI formel InChI=1S/C11H15Cl2O2PS2/c1-3-7-18-16(17,14-4-2)15-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3
LEDER CCCSP(=S)(OCC)Oc1ccc(Cl)cc1Cl
IUPAC-namn (2,4-dichlorophenoxy)-ethoxy-propylsulfanyl-sulfanylidene-λ^{5}-phosphane
Molekylvikt (g/mol) 345.25
Formel vikt 343.9628