Läs mer
Protionamide, TRC
Produktkod. 17981175
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981175 1 g
17963395 5 g
17942795 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8391001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14222-60-7
Molekylformel C9 H12 N2 S
Molekylvikt (g/mol) 180.27
Synonym 2-Propylthio-isonicotinamide, 1321TH, 2-Propyl-4-pyridinecarbothioamide, 2-Propylisonicotinylthioamide, 2-Propylthioisonicotinamide, 9778 R. P., Ektebin, Prothionamide, Protion, Protionamid, Protionamide, Protionizina, RP 9778, TH 1321, Tebeform, Trevintix, Tuberex
IUPAC-namn 2-propylpyridine-4-carbothioamide
LEDER CCCc1cc(ccn1)C(=S)N
Specifikationer
Kemiskt namn eller material Protionamide
Kvantitet 1 g
InChI formel InChI=1S/C9H12N2S/c1-2-3-8-6-7(9(10)12)4-5-11-8/h4-6H,2-3H2,1H3,(H2,10,12)
Formel vikt 180.0721