missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Protirelin acetate

Product Code. 16499308
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16499308 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499308 Supplier MedChemExpress Supplier No. HYP0002A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Protirelin Acetate is a highly conserved neuropeptide that exerts the hormonal control of thyroid-stimulating hormone (TSH) levels as well as neuromodulatory functions.

Specifications

Renhet 99.93%
CAS 120876-23-5
Innehåll och lagring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
För användning med (applikation) COVID-19-immunoregulation
Molekylvikt (g/mol) 452.46 g/mol
Produkttyp Peptides
Kvantitet 10 mg
Frakt skick Room temperature
Löslighet DMSO : 250 mg/mL (552.54 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (110.51 mM∣Need ultrasonic)
Synonym Thyrotropin-releasing-hormone acetate∣TRH acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.