missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Protocadherin Gamma Monoclonal Antibody (N159/5), FITC

Produktkod. 17878592
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17878592 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17878592 Leverantör Invitrogen™ Leverantörsnummer MA545659

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45659 was sufficient for detection of Protocadherin gamma (pan) in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins. This antibody was formerly sold as clone S159-5.

The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Protocadherin Gamma
Användningsområden Immunohistochemistry, Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona N159/5
Koncentration 1 mg/mL
Konjugera FITC
Formulering PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gen PCDHGA12
Genaccessionsnr. O60330, Q91XY4, Q9UN70, Q9UN71, Q9Y5F6, Q9Y5F7, Q9Y5F8, Q9Y5F9, Q9Y5G0, Q9Y5G1, Q9Y5G2, Q9Y5G3, Q9Y5G4, Q9Y5G5, Q9Y5G6, Q9Y5G7, Q9Y5G8, Q9Y5G9, Q9Y5H0, Q9Y5H1, Q9Y5H2, Q9Y5H3, Q9Y5H4
Gene Alias 3' RACE product; cadherin 20; cadherin 21; cadherin ME3; cadherin ME6; cadherin-20; Cadherin-21; cadherin-like 2; CDH20; CDH21; CDH-GAMMA-A5; FIB2; FIB3; fibroblast cadherin FIB2; fibroblast cadherin FIB3; fibroblast cadherin-2; Fibroblast cadherin-3; KIAA0327; KIAA0588; ME3; ME6; mKIAA0588; mKIAA4054; Pan-Gamma-Protocadherin-A; pc2c; PC43; PC-43; PCDH gamma A; Pcdh13; PCDH2; PCDHGA; PCDHGA1; PCDHGA10; PCDHGA11; PCDHGA12; Pcdhga2; PCDHGA3; Pcdhga4; PCDHGA5; PCDHGA6; PCDHGA7; Pcdhga8; PCDHGA9; PCDH-gamma-A; PCDH-GAMMA-A1; PCDH-gamma-A10; PCDH-GAMMA-A11; PCDH-gamma-A12; PCDH-GAMMA-A2; PCDH-gamma-A3; PCDH-gamma-A4; PCDH-GAMMA-A5; PCDH-gamma-A6; PCDH-GAMMA-A7; PCDH-gamma-A8; PCDH-gamma-A9; PCDH-GAMMA-B1; PCDH-GAMMA-B2; PCDH-gamma-B3; PCDH-GAMMA-B4; PCDH-GAMMA-B5; PCDH-GAMMA-B6; PCDH-gamma-B7; PCDH-GAMMA-C3; PCDH-gamma-C4; PCDH-gamma-C5; Pcdhgb1; PCDHGB2; PCDHGB3; PCDHGB4; PCDHGB5; Pcdhgb6; Pcdhgb7; PCDHGC3; Pcdhgc4; PCDHGC5; protocadherin 13; protocadherin 2; protocadherin 43; protocadherin gamma A12; protocadherin gamma B2; protocadherin gamma B6; protocadherin gamma C-VI; protocadherin gamma subfamily A, 1; protocadherin gamma subfamily A, 10; protocadherin gamma subfamily A, 11; protocadherin gamma subfamily A, 12; protocadherin gamma subfamily A, 2; protocadherin gamma subfamily A, 3; protocadherin gamma subfamily A, 4; protocadherin gamma subfamily A, 5; protocadherin gamma subfamily A, 6; protocadherin gamma subfamily A, 7; protocadherin gamma subfamily A, 8; protocadherin gamma subfamily A, 9; protocadherin gamma subfamily B, 1; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma subfamily B, 2; protocadherin gamma subfamily B, 3; protocadherin gamma subfamily B, 4; protocadherin gamma subfamily B, 5; protocadherin gamma subfamily B, 6; protocadherin gamma subfamily B, 7; protocadherin gamma subfamily C, 3; protocadherin gamma subfamily C, 4; protocadherin gamma subfamily C, 5; protocadherin gamma-A1; Protocadherin gamma-A10; protocadherin gamma-A11; Protocadherin gamma-A12; protocadherin gamma-A2; Protocadherin gamma-A3; protocadherin gamma-A4; Protocadherin gamma-A5; protocadherin gamma-A6; Protocadherin gamma-A7; protocadherin gamma-A8; Protocadherin gamma-A9; protocadherin gamma-B1; Protocadherin gamma-B2; protocadherin gamma-B3; Protocadherin gamma-B4; protocadherin gamma-B5; Protocadherin gamma-B6; protocadherin gamma-B7; Protocadherin gamma-C3; protocadherin gamma-C4; Protocadherin gamma-C5; protocadherin-2; Protocadherin-43; UNQ371/PRO707
Gensymboler Pcdhga1, Pcdhga10, Pcdhga11, PCDHGA12, Pcdhga2, PCDHGA3, PCDHGA4, Pcdhga5, PCDHGA6, Pcdhga7, PCDHGA8, Pcdhga9, PCDHGB1, PCDHGB2, PCDHGB3, Pcdhgb4, Pcdhgb5, Pcdhgb6, Pcdhgb7, Pcdhgc3, PCDHGC4, PCDHGC5
Värd art Mouse
Immunogen Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ~807-930, B subfamily amino acids ~789-912 and C subfamily amino acids ~818-941).
Reningsmetod Protein G
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 102552497, 102552705, 102552796, 116782, 252894, 252895, 252897, 26025, 291635, 291637, 364843, 364844, 498846, 498847, 498849, 498850, 5098, 553129, 56097, 56098, 56099, 56100, 56101, 56102, 56103, 56104, 56105, 56106, 56107, 56108, 56109, 56110, 56111, 56112, 56113, 56114, 641525, 641526, 680361, 680448, 8641, 93699, 93700, 93701, 93702, 93703, 93704, 93706, 93707, 93708, 93709, 93710, 93711, 93712, 93713, 93714, 93715, 93716, 93717, 93722, 93723, 93724, 9708
Målarter Human, Mouse, Rat
Innehåll och lagring 4°C, store in dark
Produkttyp Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.