Läs mer
Protocatechuic Acid Diacetate, TRC
Produktkod. 30422150
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30422150 250 mg
1 options
Produktkod. 30422150 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP838890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Protocatechuic Acid Diacetate
CAS 58534-64-8
Molekylformel C11H10O6
Kvantitet 250 mg
Synonym 3,4-Diacetoxybenzoic Acid, 3,4-Bis(acetyloxy)benzoic Acid
InChI formel InChI=1S/C11H10O6/c1-6(12)16-9-4-3-8(11(14)15)5-10(9)17-7(2)13/h3-5H,1-2H3,(H,14,15)
LEDER O=C(C1=CC=C(C(OC(C)=O)=C1)OC(C)=O)O
IUPAC-namn 3,4-diacetoxybenzoic acid
Molekylvikt (g/mol) 238.19
Formel vikt 238.05