Läs mer
Protriptyline.HCl, Lipomed
Beskrivning
Protriptyline.HCl
Specifikationer
Specifikationer
| Kemiskt namn eller material | Protriptyline HCl |
| CAS | 1225-55-4 |
| Analyt- eller komponentnamn | Protriptyline HCl |
| Frakt skick | Room Temperature |
| Molekylformel | C19 H21 N . Cl H |
| Synonym | N-Nethyl-5 H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, Concordin, MK-240, Maximed, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| LEDER | Cl.CNCCCC1c2ccccc2C=Cc3ccccc13 |
| IUPAC-namn | 3-(11 H-dibenzo[2,1-b:1',2'-e][7 ]annulen-11-yl)-N-methylpropan-1-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Visa mer |