Läs mer
Protriptyline Hydrochloride, LoGiCal
Produktkod. 30459300
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30459300 10 mg
1 options
Produktkod. 30459300 Leverantör LoGiCal Leverantörsnummer LGCFOR0979.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Protriptyline HCl
CAS 1225-55-4
Analyt- eller komponentnamn Protriptyline HCl
Frakt skick Room Temperature
Molekylformel C19 H21 N . Cl H
Certifikat och efterlevnad ISO 9001
Synonym N-Nethyl-5H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride, N-Methyl-5H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, Concordin, MK-240, Maximed, Maximet, N-Methyl-5H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, Normethyl EX4442, Protriptyline hydrochloride, Protryptyline hydrochloride, Triptil, Triptil Hydrochloride, Vivactil
InChI formel InChI=1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-10,12-13,19-20H,6,11,14H2,1H3;1H
Lösningstyp Neat
LEDER Cl.CNCCCC1c2ccccc2C=Cc3ccccc13
IUPAC-namn 3-(11H-dibenzo[2,1-b:1',2'-e][7]annulen-11-yl)-N-methylpropan-1-amine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 299.84
Kvantitet 10 mg
Formel vikt 299.1441 g/mol