missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRR14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22819-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene tethers heterochromatin to the nuclear laminar scaffold by binding heterochromatin protein 1 (HP1) and the nuclear lamina. The tether is broken during mitosis and reforms quickly after mitosis, with the encoded protein first binding HP1 and then attaching to the nuclear lamina. This protein also has been shown to promote MyoD activity and skeletal myogenesis. Two transcript variants encoding the same protein have been found for this gene.Specifications
PRR14 | |
Polyclonal | |
Unconjugated | |
PRR14 | |
proline rich 14, Proline rich protein 14, PRR14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233895, 78994 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TPN9, Q9BWN1 | |
PRR14 | |
PRR14 Fusion Protein Ag14119 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |