missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ PRR5 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 17751324
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17751324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17751324 Leverantör Invitrogen™ Leverantörsnummer PA5119923

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Positive Control: MCF-7 cell lysate, mouse liver tissue lysate, rat liver tissue lysate. Subcellular Location: Cytosol.

MARK1 (MAP/microtubule affinity-regulating kinase 1) belongs to the MARK family of serine/threonine kinases. MARK family protein kinases phosphorylate microtubule-associated proteins and may play a role in cytoskeletal stability. MARK2 refers to MAP/microtubule affinity-regulating kinase 2 isoform a [Homo sapiens]. EMK (ELKL Motif Kinase) is a small family of ser/thr protein kinases involved in the control of cell polarity, microtubule stability and cancer. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1 called MARK2. MARK3 was originally identified as a marker that was induced by treatment with DNA damaging agents, and loss of MARK3 was found with carcinogenesis in the pancreas. MARK4 contains an N terminal serine/threonine kinase domain, a central ubiquitin associated domain, and a C terminal KA1 associated kinase domain. RT PCR analysis detects upregulated expression of the gene for MARK4 in nearly all clinical hepatocellular carcinoma cells.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen PRR5
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gen PRR5
Genaccessionsnr. P85299, Q5FVG6, Q812A5
Gene Alias Arhgap8; AU043908; C030017C09Rik; C78947; FLJ20185k; PP610; proline rich 5; proline rich 5 (renal); proline-rich protein 5; Protein observed with Rictor-1; Protor1; protor-1; prr5; prr5 {ECO:0000250; Rho GTPase activating protein 8; RP1-181C9.3; UniProtKB:P85299}
Gensymboler PRR5
Värd art Rabbit
Immunogen Recombinant protein within human PRR5 aa 200-388.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 109270, 315189, 55615
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.