missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27887-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.Specifications
PRRC1 | |
Polyclonal | |
Unconjugated | |
PRRC1 | |
proline rich coiled coil 1, Protein PRRC1, PRRC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
133619, 73137 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UPH1, Q96M27 | |
Prrc1 | |
PRRC1 Fusion Protein Ag27349 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |