missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRRC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PRRC1 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825895
|
Proteintech
27887-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815895
|
Proteintech
27887-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PRRC1 gene ontology annotations related to this gene include activation of protein kinase A activity.Specifications
PRRC1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q3UPH1, Q96M27 | |
133619, 73137 | |
PRRC1 Fusion Protein Ag27349 | |
Primary | |
-20°C | |
PRRC1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
proline rich coiled coil 1, Protein PRRC1, PRRC1 | |
Prrc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |