Läs mer
Tocris Bioscience™ PRX 07034
GREENER_CHOICE
Produktkod. 18739226
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18739226 50 mg
18715406 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18739226 Leverantör Tocris Bioscience™ Leverantörsnummer 6748/50
Potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 to 8 nM)
PRX 07034 is a potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 - 8 nM). Enhances working memory in rats.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(1-(5-Chloro-2,3-dimethoxyphenyl)ethyl)-2-(methylsulfonyl)-5-(piperazin-1-yl)aniline hydrochloride
CAS 903580-39-2
Kvantitet 50 mg
Molekylformel C21H28ClN3O4S.HCl
Formel vikt 490.44
Renhet 98%