missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PRX5 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837674
|
Proteintech
17724-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16827674
|
Proteintech
17724-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The peroxiredoxin (PRX) family comprises six antioxidant proteins, PRX I, II, III, IV, V and VI, which protect cells from reactive oxygen species (ROS) by preventing the metal-catalyzed oxidation of enzymes. The PRX proteins primarily utilize thioredoxin as the electron donor for antioxidation, although they are fairly promiscuous with regard to the hydroperoxide substrate. In addition to protection from ROS, peroxiredoxins are also involved in cell proliferation, differentiation and gene expression. PRX I, II, IV and VI show diffuse cytoplasmic localization, while PRX III and V exhibit distinct mitochondrial localization. The human PRX I gene encodes a protein that is expressed in several tissues, including liver, kidney, testis, lung and nervous system. PRX II is expressed in testis, while PRX III shows expression in lung. PRX I, II and III are overexpressed in breast cancer and may be involved in its development or progression. Upregulated protein levels of PRX I and II in Alzheimer′s disease (AD) and Down syndrome (DS) indicate the involvement of PRX I and II in their pathogenesis. The human PRX IV gene is abundantly expressed in many tissues. PRX IV exists as a precursor protein, which is only detected in testis, and a processed secreted form. PRX V also exists as two forms, designated long and short. Like PRX IV, the long form of PRX V is highly expressed in testis. The short form of PRX V is more widely expressed, with high expression in liver, kidney, heart and lung. PRX VI, a 1-Cys peroxiredoxin (also known as antioxidant protein 2 or AOP2), is highly expressed in most tissues, particularly in epithelial cells. Localized to the cell cytosol, PRX VI functions independently of other peroxiredoxins and antioxidant proteins, specializing in antioxidant defense, lung phospholipid metabolism and protection of keratinocytes from cell death induced by reactive oxygen species.Specifications
PRX5 | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P30044, P99029, Q9R063 | |
113898, 25824, 54683 | |
PRX5 Fusion Protein Ag12118 | |
Primary | |
-20°C | |
PRDX5 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACR1, Alu corepressor 1, Antioxidant enzyme B166, AOEB166, AOPP, B166, Liver tissue 2D page spot 71B, Peroxiredoxin 5, Peroxiredoxin 5, mitochondrial, Peroxiredoxin V, Peroxisomal antioxidant enzyme, PLP, PMP20, PRDX5, PRDX6, Prx V, PRX5, PRXV, SBBI10, Thioredoxin peroxidase PMP20, Thioredoxin reductase, TPx type VI | |
Prdx5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |