Läs mer
Pseudo Vardenafil, TRC
Produktkod. 30008987
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008987 25 mg
30003355 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP83961525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 224788-34-5
Molekylformel C22 H29 N5 O4 S
Molekylvikt (g/mol) 459.56
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-(1-piperidinylsulfonyl)phenyl]-5-methyl-7-propyl-, Piperidine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), Piperidenafil, Pseudovardenafil
IUPAC-namn 2-(2-ethoxy-5-piperidin-1-ylsulfonylphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
LEDER CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCCCC4
Specifikationer
Kemiskt namn eller material Pseudovardenafil
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C22H29N5O4S/c1-4-9-19-23-15(3)20-22(28)24-21(25-27(19)20)17-14-16(10-11-18(17)31-5-2)32(29,30)26-12-7-6-8-13-26/h10-11,14H,4-9,12-13H2,1-3H3,(H,24,25,28)
Formel vikt 459.194
Procent renhet >95%