missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSG9 (Human) Recombinant Protein
Human PSG9 full-length ORF ( AAH20759, 35 a.a. - 426 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005678-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: EVTIEAQPPKVSEGKDVLLLVHNLPQNLPGYFWYKGEMTDLYHYIISYIVDGKIIIYGPAYSGRETVYSNASLLIQNVTRKDAGTYTLHIIKRGDETREEIRHFTFTLYLETPKPYISSSNLNPREAMEAVRLICDPETLDASYLWWMNGQSLPVTHRLQLSKTNRTLYLFGVTKYIAGPYECEIRNPVSASRSDPVTLNLLPKLPIPYITINNLNPRENKDVLAFTCEPKSENYTYIWWLNGQSLPVSPGVKRPIENRILILPSVTRNETGPYQCEIQDRYGGLRSNPVILNVLYGPDLPRIYPSFTYYRSGENLDLSCFTESNPPAEYFWTINGKFQQSGQKLFIPQITRNHSGLYACSVHNSATGKEISKSMTVKVSGPCHGDLTESQS
Specifications
AAH20759 | |
pregnancy specific beta-1-glycoprotein 9 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSG9 | |
GST |
5678 | |
Wheat germ expression system | |
10 μg | |
PSG11, PSGII | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |