Läs mer
psi-Tropine, TRC
Produktkod. 30453054
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453054 1 g
30449849 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30453054 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8926601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 135-97-7
Molekylformel C8 H15 N O
Molekylvikt (g/mol) 141.21
Synonym Pseudotropine, 3β-Tropanol, 1αH,5αH-Tropan-3β-ol (8CI), 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, exo-, Pseudotropine (6CI,7CI), (3-exo)-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol, 3-Pseudotropanol, 3β-Tropanol, NSC 43871, Pseudotropanol, ψ-Tropine
IUPAC-namn (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
LEDER CN1[C@@H]2CC[C@H]1C[C@H](O)C2
Specifikationer
Kemiskt namn eller material Pseudotropine
Kvantitet 1 g
InChI formel InChI=1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8-
Formel vikt 141.1154