missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25504-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.Specifications
PSIP1 | |
Polyclonal | |
Unconjugated | |
PSIP1 | |
CLL associated antigen KW 7, DFS 70, DFS70, LEDGF, p52, p75, PAIP, PSIP1, PSIP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11168 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75475 | |
PSIP1 | |
PSIP1 Fusion Protein Ag21886 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |