Läs mer
PSMB1 Mouse, anti-Human, Unconjugated, Clone: 1C7C5, Proteintech
Produktkod. 17231516 Handla allt Proteintech Produkter
Change view
Kvantitet:
20 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17231516 20 μL
17221516 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17231516 Leverantör Proteintech Leverantörsnummer 678471IG20UL
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Mouse Monoclonal Antibody
PSMB1(Proteasome subunit beta type-1) is also named as PSC5 and belongs to the peptidase T1B family. The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The gene encodes a 241 amino acid protein with a 28 amino acid propeptide and two glycosylation sites.
Specifikationer
Antigen PSMB1
Användningsområden Western Blot, Immunohistochemistry, ELISA
Klassificering Monoclonal
Klona 1C7C5
Konjugera Unconjugated
Värd art Mouse
Kvantitet 20 μL
Primär eller sekundär Primary
Gen-ID (Entrez) 5689
Målarter Human
Form Liquid