missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55143-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to PSMB11.
Proteasomes generate peptides that are presented by major histocompatibility complex. Three additional subunits, beta-1i, beta-2i, and beta-5i, are induced by gamma-interferon and are preferentially incorporated into proteasomes to make immunoproteasomes. PSMB11, or beta-5t, is a catalytic subunit expressed exclusively in cortical thymic epithelial cells.Specifications
PSMB11 | |
Polyclonal | |
Unconjugated | |
PSMB11 | |
beta5t, Proteasome subunit beta 5t, PSMB11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122706, 73902 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A5LHX3, Q8BG41 | |
Psmb11 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |