missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMC2 (Human) Recombinant Protein
Human PSMC2 full-length ORF ( AAH02589.1, 1 a.a. - 433 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005701-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPDYLGADQRKTKEDEKDDKPIRALDEGDIALLKTYGQSTYSRQIKQVEDDIQQLLKKINELTGIKESDTGLAPPALWDLAADKQTLQSEQPLQVARCTKIINADSEDPKYIINVKQFAKFVVDLSDQVAPTDIEEGMRVGVDRNKYQIHIPLPPKIDPTVTMMQVEEKPDVTYSDVGGCKEQIEKLREVVETPLLHPERFVNLGIEPPKGVLLFGPPGTGKTLCARAVANRTDACFIRVIGSELVQKYVGEGARMVRELFEMARTKKACLIFFDEIDAIGGARFDDGAGGDNEVQRTMLELINQLDGFDPRGNIKVLMATNRPDTLDPALMRPGRLDRKIEFSLPDLEGRTHIFKIHARSMSVERDIRFELLARLCPNSTGAEIRSVCTEAGMFAIRARRKIATEKDFLEAVNKVIKSYAKFSATPRYMTYN
Specifications
AAH02589.1 | |
proteasome (prosome, macropain) 26S subunit, ATPase, 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMC2 | |
GST |
5701 | |
Wheat germ expression system | |
25 μg | |
MGC3004, MSS1, Nbla10058, S7 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |