missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12342-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of the PA700/19S complex, which is the regulatory component of the 26S proteasome. The 26S proteosome complex is required for ubiquitin-dependent protein degradation. This protein is a non-ATPase subunit that may be involved in protein-protein interactions. Aberrant expression of this gene may paly a role in tumorigenesis. Two transcripts encoding different isoforms have been described. Pseudogenes have been identified on chromosomes 3 and 20.Specifications
PSMD10 | |
Polyclonal | |
Unconjugated | |
PSMD10 | |
Gankyrin, p28, p28(GANK), PSMD10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116722, 53380, 5716 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75832, Q9Z2X2, Q9Z2X3 | |
Psmd10 | |
PSMD10 Fusion Protein Ag2994 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |