missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMD13 (Human) Recombinant Protein
Human PSMD13 full-length ORF ( AAH01747, 1 a.a. - 376 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005719-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKDVPGFLQQSQSSGPGQPAVWHRLEELYTKKLWHQLTLQVLDFVQDPCFAQGDGLIKLYENFISEFEHRVNPLSLVEIILHVVRQMTDPNVALTFLEKTREKVKSSDEAVILCKTAIGALKLNIGDLQVTKETIEDVEEMLNNLPGVTSVHSRFYDLSSKYYQTIGNHASYYKDALRFLGCVDIKDLPVSEQQERAFTLGLAGLLGEGVFNFGELLMHPVLESLRNTDRQWLIDTLYAFNSGNVERFQTLKTAWGQQPDLAANEAQLLRKIQLLCLMEMTFTRPANHRQLTFEEIAKSAKITVNEVELLVMKALSVGLVKGSIDEVDKRVHMTWVQPRVLDLQQIKGMKDRLEFWCTDVKSMEMLVEHQAHDILT
Specifications
AAH01747 | |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 13 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMD13 | |
GST |
5719 | |
Wheat germ expression system | |
25 μg | |
HSPC027, Rpn9, S11, p40.5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |