missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMD3 (Human) Recombinant Protein
Human PSMD3 full-length ORF ( AAH20518.1, 1 a.a. - 534 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005709-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKQEGSARRRGADKAKPPPGGGEQEPPPPPAPQDVEMKEEAATGGGSTGEADGKTAAAAAEHSQRELDTVTLEDIKEHVKQLEKAVSGKEPRFVLRALRMLPSTSRRLNHYVLYKAVQGFFTSNNATRDFLLPFLEEPMDTEADLQFRPRTGKAASTPLLPEVEAYLQLLVVIFMMNSKRYKEAQKISDDLMQKISTQNRRALDLVAAKCYYYHARVYEFLDKLDVVRSFLHARLRTATLRHDADGQATLLNLLLRNYLHYSLYDQAEKLVSKSVFPEQANNNEWARYLYYTGRIKAIQLEYSEARRTMTNALRKAPQHTAVGFKQTVHKLLIVVELLLGEIPDRLQFRQPSLKRSLMPYFLLTQAVRTGNLAKFNQVLDQFGEKFQADGTYTLIIRLRHNVIKTGVRMISLSYSRISLADIAQKLQLDSPEDAEFIVAKAIRDGVIEASINHEKGYVQSKEMIDIYSTREPQLAFHQRISFCLDIHNMSVKAMRFPPKSYNKDLESAEERREREQQDLEFAKEMAEDDDDSFP
Specifications
AAH20518.1 | |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMD3 | |
GST |
5709 | |
Wheat germ expression system | |
10 μg | |
P58, RPN3, S3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |