missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD4 Mouse anti-Human, Clone: 3G1A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66179-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PSMD4.
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.Specifications
PSMD4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55036 | |
PSMD4 | |
PSMD4 Fusion Protein Ag7010 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3G1A1 | |
Unconjugated | |
PSMD4 | |
AF, AF 1, Antisecretory factor 1, ASF, MCB1, PSMD4, pUB R5, Rpn10, S5A | |
Mouse | |
Protein G | |
RUO | |
5710 | |
-20°C | |
Liquid |