missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11469-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PSMD5.
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base.Specifications
PSMD5 | |
Polyclonal | |
Unconjugated | |
PSMD5 | |
26S protease subunit S5 basic, 26S proteasome subunit S5B, KIAA0072, PSMD5, S5B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296651, 5711, 66998 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16401, Q8BJY1 | |
PSMD5 | |
PSMD5 Fusion Protein Ag2026 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |