missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMD8 (Human) Recombinant Protein
Human PSMD8 full-length ORF ( NP_002803, 1 a.a. - 257 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_002803 |
---|---|
Gene ID (Entrez) | 5714 |
Name | proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123675
|
Abnova™
H00005714-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16133675
|
Abnova™
H00005714-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MYEQLKGEWNRKSPNLSKCGEELGRLKLVLLELNFLPTTGTKLTKQQLILARDILEIGAQWSILRKDIPSFERYMAQLKCYYFDYKEQLPESAYMHQLLGLNLLFLLSQNRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQLEMIV
Specifications
NP_002803 | |
proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
HIP6, HYPF, MGC1660, Nin1p, Rpn12, S14, p31 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5714 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMD8 | |
GST |