missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD9 Mouse anti-Human, Unconjugated, Clone: 1H2G1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PSMD9 |
---|---|
Clone | 1H2G1 |
Concentration | 940 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849915
|
Proteintech
67338-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839915
|
Proteintech
67338-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The PSMD9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMD9. This antibody recognizes human, mouse, rat antigen. The PSMD9 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PSMD9 | |
940 μg/mL | |
Monoclonal | |
Mouse | |
5715 | |
Primary |
1H2G1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |