missing translation for 'onlineSavingsMsg'
Learn More

PSME3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14907-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823814

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
Specifications

Specifications

PSME3
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61289, P61290
Ki, Ki nuclear autoantigen, PA28 gamma, PA28G, PA28gamma, PSME3, REG GAMMA
Rabbit
IgG
20 μL
-20°C
Primary
10197, 19192, 287716
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PSME3
Liquid
Psme3
PSME3 Fusion Protein Ag6702
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers