Learn More
DWK Life Sciences DURAN™ GLS 80™ High Temperature Screw Cap, with PTFE-Coated Silicone Cap Liner
1572.25 SEK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
White
Description
The material used for this GLS 80 closure is a special compound based on polyarylsulphone. As a result, the chemical, thermal and mechanical properties of the material are improved compared to polypropylene. Thanks to the shortened triple thread, the DURAN™ GLS 80™ bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: Platinum-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.
- PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components
- Screw cap seal with PTFE faced cap liner for efficient sealing
- The matching PTFE pouring ring permits drip-free pouring and clean, safe working
Specifications
Diameter (Metric) | 88.5 mm |
Description | Screw cap |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping |
Material | PSU, PBT, ETFE |
Quantity | 5/Pk. |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |