missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PTCH1 (Human) Recombinant Protein
Human PTCH1 partial ORF (NP_000255.2, 651 a.a. - 750 a.a.) recombinant protein with GST tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_000255.2 |
---|---|
Gene ID (Entrez) | 5727 |
Name | patched homolog 1 (Drosophila) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173725
|
Abnova™
H00005727-Q02.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183725
|
Abnova™
H00005727-Q02.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: AHETQITMQSTVQLRTEYDPHTHVYYTTAEPRSEISVQPVTVTQDTLSCQSPESTSSTRDLLSQFSDSSLHCLEPPCTKWTLSSFAEKHYAPFLLKPKAK
Specifications
NP_000255.2 | |
patched homolog 1 (Drosophila) | |
125% SDS-PAGE Stained with Coomassie Blue | |
BCNS, FLJ26746, FLJ42602, HPE7, NBCCS, PTC, PTC1, PTCH, PTCH11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5727 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PTCH1 | |
GST |