Learn More
PTEN Antibody, R&D Systems™
Product Code. 18609965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. Quantity Unit Size
18609965 0.1 mg 0.1mg
1 options
Product Code. 18609965 Supplier Novus Biologicals Supplier No. NBP2445380.1mg
 Restricted Item
Beräknad leverans: 27-11-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
PTEN Polyclonal antibody specifically detects PTEN in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen PTEN
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 μg/mL, Flow Cytometry 0.5 - 1 μg/million cells in 0.1 mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 1 - 2 μg/mL
Formulation 10mM PBS and 0.05% BSA
Gene Accession No. P60484
Gene Alias BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM
Host Species Rabbit
Immunogen Recombinant full-length human PTEN protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Lipid and Metabolism, Metastasis, mTOR Pathway, Phospho Specific, Protein Phosphatase, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5728
Target Species Human, Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus