Läs mer
Pterine-6-carboxylic Acid, TRC
Produktkod. 30136691
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136691 1 g
30005295 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8398801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 948-60-7
Molekylformel C7 H5 N5 O3
Molekylvikt (g/mol) 207.15
Synonym 6-Pteridinecarboxylic acid, 2-amino-3,4-dihydro-4-oxo-, 6-Pteridinecarboxylic acid, 2-amino-1,4-dihydro-4-oxo- (8CI,9CI), 6-Pteridinecarboxylic acid, 2-amino-4-hydroxy- (6CI), 2-Amino-3,4-dihydro-4-oxo-6-pteridinecarboxylic acid, 2-Amino-4-hydroxy-6-carboxypteridine, 2-Amino-4-hydroxypteridine-6-carboxylic acid, 2-Amino-4-hydroxypterin-6-carboxylic acid, 2-Amino-6-carboxy-4-hydroxypteridine, 6-Carboxypterin, NSC 96893, Pterin-6-carboxylic acid, Ranachrome 5, Pterine-6-carboxylic Acid
IUPAC-namn 2-amino-4-oxo-3H-pteridine-6-carboxylic acid
LEDER NC1=Nc2ncc(nc2C(=O)N1)C(=O)O
Specifikationer
Kemiskt namn eller material Pterine-6-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C7H5N5O3/c8-7-11-4-3(5(13)12-7)10-2(1-9-4)6(14)15/h1H,(H,14,15)(H3,8,9,11,12,13)
Formel vikt 207.0392
Procent renhet >95%