Läs mer
Pteroic Acid (>85%), TRC
Produktkod. 30135982
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135982 50 mg
30005059 100 mg
30011736 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135982 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP84011050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119-24-4
Molekylformel C14 H12 N6 O3
Molekylvikt (g/mol) 312.28
Synonym 4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino]benzoic Acid, Folic Acid Imp. D (EP), Pteroic Acid
IUPAC-namn 4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoic acid
LEDER NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)O)cnc2N1
Specifikationer
Kemiskt namn eller material Pteroic acid
Kvantitet 50 mg
InChI formel InChI=1S/C14H12N6O3/c15-14-19-11-10(12(21)20-14)18-9(6-17-11)5-16-8-3-1-7(2-4-8)13(22)23/h1-4,6,16H,5H2,(H,22,23)(H3,15,17,19,20,21)
Formel vikt 312.0971