Läs mer
Pteroyltri-Gamma-L-glutamic acid (>85%), TRC
Produktkod. 30422937
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30422937 1 mg
1 options
Produktkod. 30422937 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8402201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pteroyltri-Gamma-L-glutamic acid
CAS 89-38-3
Molekylformel C29 H33 N9 O12
Kvantitet 1 mg
Synonym Glutamic acid','N-[N-(N-pteroyl-L-γ-glutamyl)-L-γ-glutamyl]-','L- (6CI,7CI,8CI)
InChI formel InChI=1S/C29H33N9O12/c30-29-37-23-22(25(44)38-29)33-15(12-32-23)11-31-14-3-1-13(2-4-14)24(43)36-18(28(49)50)6-9-20(40)34-16(26(45)46)5-8-19(39)35-17(27(47)48)7-10-21(41)42/h1-4,12,16-18,31H,5-11H2,(H,34,40)(H,35,39)(H,36,43)(H,41,42)(H,45,46)(H,47,48)(H,49,50)(H3,30,32,37,38,44)/t16-,17-,18-/m0/s1
LEDER NC1=Nc2ncc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)N[C@@H](CCC(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)O)C(=O)O)nc2C(=O)N1
IUPAC-namn (2S)-2-[[(4S)-4-[[(4S)-4-[[4-[(2-amino-4-oxo-3H-pteridin-6-yl)methylamino]benzoyl]amino]-4-carboxybutanoyl]amino]-4-carboxybutanoyl]amino]pentanedioic acid
Molekylvikt (g/mol) 699.625
Formel vikt 699.225