missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Blade for Plain Shafts

Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals. Cowie Technology™ PTFE Blade for Plain Shafts fit all standard shafts for blades.

Brand:  Fisherbrand™

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10036031

  • 37.64 SEK / Each


Estimated Shipment
14-01-2019
Add to basket

Description

Description

  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications

Specifications

Blades for Plain Shafts
Stirring in Narrow Neck Vessel
PTFE
3mm
9g
1Each
PTFE Shaft Stirrer, Plain Shaft for Blades
-200 to +280°C
75mm
25mm
Videos
View accessory products