Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, straight
For safe connection of hoses.
890.00 SEK - 1855.00 SEK
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10376272
|
DWK Life Sciences
286230005 |
GL 14 |
890.00 SEK
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10561293
|
DWK Life Sciences
286230202 |
GL 18 |
1025.00 SEK
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15273428
|
DWK Life Sciences
286230408 |
GL 25 |
1855.00 SEK
Pack of 10 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
- Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
10/Pk. | |
Straight |