Learn More
Cole-Parmer™ PTFE Manifold
Organize your fluid paths with these thermoplastic manifolds
3705.00 SEK - 4305.00 SEK
Specifications
Material | PTFE |
---|---|
Min. Operating Temperature (English) | -200°F |
Min. Operating Temperature (Metric) | -129°C |
Inlet | 1/2 in. |
Width (Metric) | 3.81 cm |
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Quantity | Price | Quantity & Availability | |||||
11795268
|
Cole-Parmer™
31521-23 |
1/Pk |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11725278
|
Cole-Parmer™
31521-29 |
1/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Chemically-inert PTFE handles the most aggressive chemical solutions.
- Available in a variety of configurations to suit a wide range of applications.
Specifications
PTFE | |
-129°C | |
3.81 cm | |
120 psi | |
150°C |
-200°F | |
1/2 in. | |
3.81 cm | |
300°F |